• USA Home
  • Search Results

  • toluenesolution1234510888311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Toluene solution

2 Products
Synonym:   Toluene solution
CAS Number:
 EC Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Special Grade
  Purity Grade
  Package quantity
  Concentration
Show More