• USA Home
  • Search Results

  • topiramateimpuritye180165748711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Topiramate impurity E

1 Products
Synonym:   D-(−)-Fructose, D-Levulose, Fruit sugar
Image of Topiramate impurity E
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1239004
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More