• USA Home
  • Search Results

  • trans11eicosenoicacid310516232284311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-11-Eicosenoic acid

1 Products
Image of <I>trans</I>-11-Eicosenoic acid
CAS Number:
Empirical Formula (Hill Notation):
 C20H38O2
Molecular Weight:
 310.51
 Beilstein/REAXYS Number:
 1727314
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More