• USA Home
  • Search Results

  • trans12diaminocyclohexane11419112122811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(±)-trans-1,2-Diaminocyclohexane

1 Products
Synonym:   (±)-trans-1,2-Cyclohexanediamine
Image of (±)-<I>trans</I>-<WBR>1,2-<WBR>Diaminocyclohexane
CAS Number:
Linear Formula:
 C6H10(NH2)2
Molecular Weight:
 114.19
 Beilstein/REAXYS Number:
 3193807
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More