• USA Home
  • Search Results

  • trans2cis6nonadienal1382155748211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-2,cis-6-Nonadienal

3 Products
Synonym:   (E,Z)-2,6-Nonadienal, Violet leaf aldehyde
Image of <I>trans</I>-2,<I>cis</I>-6-Nonadienal
CAS Number:
Linear Formula:
 C2H5CH=CHCH2CH2CH=CHCHO
Molecular Weight:
 138.21
 Council of Europe no.:
 659
 EC Number:
 FEMA Number:
 3377
 Flavis number:
 5.058
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Agency/Method
  Assay
Show More