• USA Home
  • Search Results

  • trans2decenal15425391381311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-2-Decenal

3 Products
Image of <I>trans</I>-2-Decenal
CAS Number:
Linear Formula:
 CH3(CH2)6CH=CHCHO
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 1704445
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More