• USA Home
  • Search Results

  • trans2hexen1al9814672826311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-2-Hexen-1-al

4 Products
Image of <I>trans</I>-2-Hexen-1-al
CAS Number:
Linear Formula:
 CH3CH2CH2CH=CHCHO
Molecular Weight:
 98.14
 Beilstein/REAXYS Number:
 1699684
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Document Links
  Allergen
  Organoleptic property
  Assay
  Agency/Method
Show More