• USA Home
  • Search Results

  • trans2hexenoicacid114141341969711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-2-Hexenoic acid

2 Products
Image of <I>trans</I>-2-Hexenoic acid
CAS Number:
Linear Formula:
 CH3CH2CH2CH=CHCO2H
Molecular Weight:
 114.14
 Beilstein Registry Number:
 1720443
 Council of Europe no.:
 11777
 EC Number:
 FEMA Number:
 3169
 Flavis number:
 8.054
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More