• USA Home
  • Search Results

  • trans3hexen1ol1001692897211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-3-Hexen-1-ol

2 Products
Synonym:   trans-3-Hexenol
Image of <I>trans</I>-3-Hexen-1-ol
CAS Number:
Linear Formula:
 C2H5CH=CHCH2CH2OH
Molecular Weight:
 100.16
 Beilstein/REAXYS Number:
 1719713
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More