• USA Home
  • Search Results

  • transaconiticacid17411402365811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-Aconitic acid

2 Products
Synonym:   trans-Propene-1,2,3-tricarboxylic acid
Image of <I>trans</I>-Aconitic acid
CAS Number:
Linear Formula:
 HO2CCH2C(CO2H)=CHCO2H
Molecular Weight:
 174.11
 Beilstein/REAXYS Number:
 1725830
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More