• USA Home
  • Search Results

  • transcaffeicacid1801650116611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-Caffeic acid

2 Products
Synonym:   trans-3,4-Dihydroxycinnamic acid
Image of <I>trans</I>-Caffeic acid
CAS Number:
Empirical Formula (Hill Notation):
 C9H8O4
Molecular Weight:
 180.16
 Beilstein Registry Number:
 1954563
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More