• USA Home
  • Search Results

  • transcinnamicacid1481614010311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-Cinnamic acid

6 Products
Synonym:   trans-3-Phenylacrylic acid, Cinnamic acid
Image of <I>trans</I>-Cinnamic acid
CAS Number:
Linear Formula:
 C6H5CH=CHCOOH
Molecular Weight:
 148.16
 Beilstein/REAXYS Number:
 1905952
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Assay
  Agency/Method
Show More