• USA Home
  • Search Results

  • transfluthrin3711511871289311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Transfluthrin

1 Products
Image of Transfluthrin
CAS Number:
Empirical Formula (Hill Notation):
 C15H12Cl2F4O2
Molecular Weight:
 371.15
 EC Number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More