• USA Home
  • Search Results

  • transglutaconicacid1301062848811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans-Glutaconic acid

1 Products
Image of <I>trans</I>-Glutaconic acid
CAS Number:
Linear Formula:
 HOOCCH2CH=CHCOOH
Molecular Weight:
 130.10
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More