• USA Home
  • Search Results

  • transtransdibenzylideneacetone234293522579711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

trans,trans-Dibenzylideneacetone

1 Products
Synonym:   1,5-Diphenyl-1,4-pentadien-3-one, trans,trans-1,5-Diphenyl-1,4-pentadien-3-one
Image of <I>trans</I>,<I>trans</I>-Dibenzylideneacetone
CAS Number:
Linear Formula:
 C6H5CH=CHCOCH=CHC6H5
Molecular Weight:
 234.29
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More