• USA Home
  • Search Results

  • trazodonehydrochloride408322533239211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trazodone hydrochloride

3 Products
Synonym:   2-[3-[4-(3-Chlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one hydrochloride
Image of Trazodone hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C19H22ClN5O · HCl
Molecular Weight:
 408.32
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  Manufacturer designation
Show More