• USA Home
  • Search Results

  • tretinoin3004430279411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tretinoin

3 Products
Synonym:   ATRA, Retinoic acid, Tretinoin, Vitamin A acid, all-trans-Retinoic acid
Image of Tretinoin
CAS Number:
Empirical Formula (Hill Notation):
 C20H28O2
Molecular Weight:
 300.44
 Beilstein/REAXYS Number:
 2057223
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Manufacturer designation
  Anal. Standard Form
  Physical Form
Show More