• USA Home
  • Search Results

  • triacetin2182010276111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triacetin

7 Products
Synonym:   1,2,3-Triacetoxypropane, 1,2,3-Triacetylglycerol, Glyceryl triacetate
Image of Triacetin
CAS Number:
Linear Formula:
 (CH3COOCH2)2CHOCOCH3
Molecular Weight:
 218.20
 Beilstein/REAXYS Number:
 1792353
 E Number:
 E 1518
 EC Number:
 FEMA Number:
 2007
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Potency
  Biological Source
  Anal. Standard Form
  Physical Form
  Application(s)
  Assay
  Agency/Method
Show More