• USA Home
  • Search Results

  • triamterene2532639601011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triamterene

5 Products
Synonym:   2,4,7-Triamino-6-phenylpteridine, 6-Phenyl-2,4,7-pteridinetriamine
Image of Triamterene
CAS Number:
Empirical Formula (Hill Notation):
 C12H11N7
Molecular Weight:
 253.26
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
  Special Grade
  Anal. Standard Form
Show More