• USA Home
  • Search Results

  • tribenzylphosphine30437765089711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tribenzylphosphine

1 Products
Image of Tribenzylphosphine
CAS Number:
Linear Formula:
 (C6H5CH2)3P
Molecular Weight:
 304.37
 Beilstein Registry Number:
 650179
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
336947 Tribenzylphosphine, 98%,