• USA Home
  • Search Results

  • tribromoaceticacid296747596711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tribromoacetic acid

1 Products
Image of Tribromoacetic acid
CAS Number:
Linear Formula:
 CBr3CO2H
Molecular Weight:
 296.74
 Beilstein/REAXYS Number:
 1755150
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
T48208 Tribromoacetic acid, 99%,