• USA Home
  • Search Results

  • tributyl2acetylcitrate402487790711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributyl 2-acetylcitrate

2 Products
Synonym:   Acetyl tributyl citrate
Image of Tributyl 2-acetylcitrate
CAS Number:
Linear Formula:
 CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
Molecular Weight:
 402.48
 Beilstein/REAXYS Number:
 2303316
 EC Number:
 FEMA Number:
 3080
 Flavis number:
 9.511
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Agency/Method
Show More