• USA Home
  • Search Results

  • tributylamine1853510282911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributylamine

4 Products
Image of Tributylamine
CAS Number:
Linear Formula:
 (CH3CH2CH2CH2)3N
Molecular Weight:
 185.35
 EC Index Number
 203-058-7
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Purity Grade
Show More