• USA Home
  • Search Results

  • tributylcitrate360447794111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributyl citrate

4 Products
Image of Tributyl citrate
CAS Number:
Empirical Formula (Hill Notation):
 C18H32O7
Molecular Weight:
 360.44
 Beilstein/REAXYS Number:
 1806072
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Assay
Show More