• USA Home
  • Search Results

  • tributylphosphate2663112673811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributyl phosphate

7 Products
Synonym:   TBP, TBPA
Image of Tributyl phosphate
CAS Number:
Linear Formula:
 (CH3(CH2)3O)3PO
Molecular Weight:
 266.31
 Beilstein/REAXYS Number:
 1710584
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Potency
  Application(s)
  Special Grade
  Solubility
Show More