• USA Home
  • Search Results

  • tributylphosphine2023299840311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributylphosphine

2 Products
Synonym:   P(n-Bu)3, TBP, Tris(trimethylsilyl)phosphane
Image of Tributylphosphine
CAS Number:
Linear Formula:
 [CH3(CH2)3]3P
Molecular Weight:
 202.32
 Beilstein Registry Number:
 1738261
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
  Assay
Show More