• USA Home
  • Search Results

  • tributylphosphinesolution2023299840311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributylphosphine solution

1 Products
Synonym:   TBP
Image of Tributylphosphine solution
CAS Number:
Empirical Formula (Hill Notation):
 C12H27P
Molecular Weight:
 202.32
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More