• USA Home
  • Search Results

  • tributyltinchloride32551146122911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tributyltin chloride

3 Products
Synonym:   TBTC, Tributylchlorotin
Image of Tributyltin chloride
CAS Number:
Linear Formula:
 [CH3(CH2)3]3SnCl
Molecular Weight:
 325.51
 Beilstein/REAXYS Number:
 3535715
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More