• USA Home
  • Search Results

  • trichloroaceticacidsolution163397603911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trichloroacetic acid solution

1 Products
Synonym:   TCA
Image of Trichloroacetic acid solution
CAS Number:
Linear Formula:
 Cl3CCOOH
Molecular Weight:
 163.39
 Beilstein/REAXYS Number:
 970119
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Application(s)
  Physical Form
Show More