• USA Home
  • Search Results

  • trichlorofluoromethanesolution123457569411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trichlorofluoromethane solution

2 Products
Image of Trichlorofluoromethane solution
CAS Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Chemical Composition
  Package quantity
  Composite Dimension
  Anal. Standard Form
  Special Grade
  Application(s)
Show More