• USA Home
  • Search Results

  • trichostatina302375888019611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trichostatin A

1 Products
Synonym:   TSA, [R-(E,E)]-7-[4-(Dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxo-2,4-heptadienamide
Image of Trichostatin A
CAS Number:
Empirical Formula (Hill Notation):
 C17H22N2O3
Molecular Weight:
 302.37
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Biological Source
  Assay
Show More