• USA Home
  • Search Results

  • tricine17917570404111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Tricine

5 Products
Synonym:   N-[Tris(hydroxymethyl)methyl]glycine, Tricine
Image of Tricine
CAS Number:
Linear Formula:
 (HOCH2)3CNHCH2CO2H
Molecular Weight:
 179.17
 Beilstein/REAXYS Number:
 1937804
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Solubility
  Product Line
  Melting Point
  Special Grade
Show More