• USA Home
  • Search Results

  • triethyl2acetylcitrate318327789411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triethyl 2-acetylcitrate

2 Products
Synonym:   Acetyl triethyl citrate
Image of Triethyl 2-acetylcitrate
CAS Number:
Linear Formula:
 CH3COOC(COOCH2CH3)(CH2COOCH2CH3)2
Molecular Weight:
 318.32
 Beilstein/REAXYS Number:
 1804947
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More