• USA Home
  • Search Results

  • triethyld15amine116286668879711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triethyl-d15-amine

1 Products
Image of Triethyl-d<SUB>15</SUB>-amine
CAS Number:
Linear Formula:
 (C2D5)3N
Molecular Weight:
 116.28
 MDL number:
 NACRES:
 NA.12
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More