• USA Home
  • Search Results

  • triethylenetetramine1462311224311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triethylenetetramine

3 Products
Synonym:   DEH 24, EPH 925, TETA
Image of Triethylenetetramine
CAS Number:
Linear Formula:
 NH2CH2CH2(NHCH2CH2)2NH2
Molecular Weight:
 146.23
 Beilstein/REAXYS Number:
 605448
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Solubility
Show More