• USA Home
  • Search Results

  • triethyloxoniumtetrafluoroboratesolution1899936839811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triethyloxonium tetrafluoroborate solution

1 Products
Image of Triethyloxonium tetrafluoroborate solution
CAS Number:
Linear Formula:
 (C2H5)3O(BF4)
Molecular Weight:
 189.99
 Beilstein/REAXYS Number:
 3598090
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More