• USA Home
  • Search Results

  • triethylphosphinesolution1181655470111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triethylphosphine solution

3 Products
Synonym:   PEt3, Triethylphosphorous solution
Image of Triethylphosphine solution
CAS Number:
Linear Formula:
 (C2H5)3P
Molecular Weight:
 118.16
 Beilstein/REAXYS Number:
 969170
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Reaction Suitability
  Density
Show More