• USA Home
  • Search Results

  • trifluoroaceticacid114027605111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trifluoroacetic acid

13 Products
Synonym:   TFA
Image of Trifluoroacetic acid
CAS Number:
Linear Formula:
 CF3COOH
Molecular Weight:
 114.02
 Beilstein/REAXYS Number:
 742035
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Purity Grade
  Ice Shipping
  Density
  Suitability
  Product Line
  UV-Absorption Condition
  Application(s)
  Special Grade
  pH Value
  Physical Form
  Solubility
  Assay
Show More

Product #
Description
302031 Trifluoroacetic acid, for HPLC, ≥99.0%,
T6508 Trifluoroacetic acid, ReagentPlus®, 99%,
91707 Trifluoroacetic acid, puriss. p.a., for HPLC, ≥99.0% (GC),
8.08260 Trifluoroacetic acid, for synthesis,
80457 Trifluoroacetic acid, eluent additive for LC-MS, LiChropur, ≥99.0% (GC),
1.08262 Trifluoroacetic acid, for spectroscopy Uvasol®,
299537 Trifluoroacetic acid, ≥99%, for protein sequencing,
TX1276 Trifluoroacetic acid, HPLC,
74564 Trifluoroacetic acid, analytical standard,
1.08178 Trifluoroacetic acid, for protein sequence analysis,
900518 Trifluoroacetic acid, anhydrous, ZerO2®, ≥99.0%,
1.08218 Trifluoroacetic acid, (25% solution in water) for protein sequenation,
TX1275 Trifluoroacetic acid, Meets Reagent Specifications for testing USP/NF monographs,