• USA Home
  • Search Results

  • trifluoromethanesulfonicacid15008149313611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trifluoromethanesulfonic acid

3 Products
Synonym:   Triflic acid
Image of Trifluoromethanesulfonic acid
CAS Number:
Linear Formula:
 CF3SO3H
Molecular Weight:
 150.08
 Beilstein/REAXYS Number:
 1812100
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More