• USA Home
  • Search Results

  • trigalacturonicacid54639603745211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trigalacturonic acid

1 Products
Synonym:   α-D-GalA-(1→4)-α-D-GalA-(1→4)-α-D-GalA, α-D-GalA-(1→4)-α-D-GalA-(1→4)-D-GalA
Image of Trigalacturonic acid
CAS Number:
Empirical Formula (Hill Notation):
 C18H26O19
Molecular Weight:
 546.39
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More