• USA Home
  • Search Results

  • triirondodecacarbonyl503661768552811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triirondodecacarbonyl

1 Products
Synonym:   Dodecacarbonyltriiron, tri-Irondodecacarbonyl
Image of Triirondodecacarbonyl
CAS Number:
Linear Formula:
 Fe3(CO)12
Molecular Weight:
 503.66
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More