• USA Home
  • Search Results

  • triisobutylaluminumsolution1983210099211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triisobutylaluminum solution

2 Products
Synonym:   TIBA solution
Image of Triisobutylaluminum solution
CAS Number:
Linear Formula:
 [(CH3)2CHCH2]3Al
Molecular Weight:
 198.32
 Beilstein/REAXYS Number:
 3587328
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More