• USA Home
  • Search Results

  • triisopropylsilane15836648579611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Triisopropylsilane

2 Products
Image of Triisopropylsilane
CAS Number:
Linear Formula:
 ((CH3)2CH)3SiH
Molecular Weight:
 158.36
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More