• USA Home
  • Search Results

  • triisopropylsilylacetylene182388934306611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Triisopropylsilyl)acetylene

1 Products
Synonym:   Ethynyltriisopropylsilane
Image of (Triisopropylsilyl)<WBR>acetylene
CAS Number:
Linear Formula:
 [(CH3)2CH]3SiC≡CH
Molecular Weight:
 182.38
 Beilstein/REAXYS Number:
 3536241
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More