• USA Home
  • Search Results

  • trimethylaminehydrochloride955759381711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethylamine hydrochloride

2 Products
Synonym:   Trimethylammonium chloride
Image of Trimethylamine hydrochloride
CAS Number:
Linear Formula:
 (CH3)3N · HCl
Molecular Weight:
 95.57
 Beilstein/REAXYS Number:
 3905063
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Titertest
Show More