• USA Home
  • Search Results

  • trimethylaminenoxide7511118478711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethylamine N-oxide

1 Products
Synonym:   TMANO
Image of Trimethylamine <I>N</I>-oxide
CAS Number:
Linear Formula:
 (CH3)3N(O)
Molecular Weight:
 75.11
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More