• USA Home
  • Search Results

  • trimethyleneoxide580850330011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethylene oxide

1 Products
Synonym:   1,3-Epoxypropane, 1,3-Propylene oxide, Cyclooxabutane, Oxacyclobutane, Oxetane
Image of Trimethylene oxide
CAS Number:
Empirical Formula (Hill Notation):
 C3H6O
Molecular Weight:
 58.08
 Beilstein/REAXYS Number:
 102382
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
Show More

Product #
Description
T76201 Trimethylene oxide, 97%,