• USA Home
  • Search Results

  • trimethylphosphate1400751256111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethyl phosphate

3 Products
Synonym:   TMP, TMPA, TMPO
Image of Trimethyl phosphate
CAS Number:
Linear Formula:
 (CH3O)3PO
Molecular Weight:
 140.07
 Beilstein/REAXYS Number:
 1071731
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
Show More