• USA Home
  • Search Results

  • trimethylphosphine760859409211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethylphosphine

1 Products
Synonym:   PMe3
Image of Trimethylphosphine
CAS Number:
Linear Formula:
 (CH3)3P
Molecular Weight:
 76.08
 Beilstein Registry Number:
 969138
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More

Product #
Description
323322 Trimethylphosphine, 97%,