• USA Home
  • Search Results

  • trimethylpropargylsilane112241336164311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Trimethyl(propargyl)silane

1 Products
Synonym:   2-Propynyl-trimethylsilane, 3-(Trimethylsilyl)propyne
Image of Trimethyl(propargyl)<WBR>silane
CAS Number:
Linear Formula:
 HC≡CCH2Si(CH3)3
Molecular Weight:
 112.24
 Beilstein/REAXYS Number:
 2036316
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Additive
Show More